siRNA Details
| Gene Symbol: Epb4.1l4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019427.2 | NP_062300.2 | 1 | 262 | ||
| GenBank mRNA | AB032366.1 | - | 1 | 757 | ||
| AK144315.1 | - | 1 | 258 | |||
| AK165783.1 | - | 1 | 255 | |||
| BB612044.1 | - | 1 | 263 | |||
| BC113123.1 | - | 2 | 106 | |||

Epb4.1l4b
Genomic Map