siRNA Details
| Gene Symbol: Epb4.1l4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB032366.1 | - | 7 | 1142 | ||
| BC113123.1 | - | 8 | 491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epb4.1l4b| Gene Symbol: Epb4.1l4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB032366.1 | - | 7 | 1142 | ||
| BC113123.1 | - | 8 | 491 | |||