siRNA Details
| Gene Symbol: Rfk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019437.3 | NP_062310.1 | 4 | 1992 | ||
| GenBank mRNA | AK002806.1 | - | 4 | 1953 | ||
| BB758770.1 | - | 1 | 49 | |||
| BC033521.1 | - | 4 | 1894 | 4527980 | ||
| BC051021.1 | - | 4 | 1903 | 6315425 | ||

Rfk
Genomic Map