siRNA Details
| Gene Symbol: Gabbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019439.3 | NP_062312.3 | 5, 6 | 828 | ||
| GenBank mRNA | AF008649.1 | - | 5, 6 | 712 | ||
| AF114168.2 | - | 5, 6 | 726 | |||
| AF120255.1 | - | 1 | 156 | |||
| AK156397.1 | - | 4, 5 | 713 | |||
| BC054735.1 | - | 5, 6 | 828 | 6401243 | ||
| BC056990.1 | - | 1 | 205 | 6821442 | ||
More Information
Other Species Targeted
Set Membership: |
|

Gabbr1
Genomic Map