siRNA Details
| Gene Symbol: Irgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019440.3 | NP_062313.3 | 2 | 892 | ||
| GenBank mRNA | AJ007972.1 | - | 1 | 693 | ||
| AK142291.1 | - | 2 | 894 | |||
| AK152870.1 | - | 2 | 833 | |||
| AK153309.1 | - | 2 | 833 | |||
| AK165347.1 | - | 2 | 830 | |||

Irgm2
Genomic Map