siRNA Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018388.1 | - | 8 | 1048 | 4209296 | |
| BC023731.1 | - | 2 | 506 | 5352340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc93b1| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018388.1 | - | 8 | 1048 | 4209296 | |
| BC023731.1 | - | 2 | 506 | 5352340 | ||