siRNA Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019454.3 | NP_062327.2 | 5 | 1069 | ||
| GenBank mRNA | AB043893.1 | - | 5 | 712 | ||
| AF253469.1 | - | 5 | 1024 | |||
| AF273454.1 | - | 5 | 1090 | |||
| AK004739.1 | - | 5 | 1037 | |||
| AK031699.1 | - | 5 | 1038 | |||
| AK052322.1 | - | 5 | 1070 | |||
| AK165517.1 | - | 5 | 1038 | |||
| AK171717.1 | - | 5 | 1036 | |||
| BC042497.1 | - | 5 | 1024 | 4017786 | ||
| BC049130.1 | - | 5 | 1029 | 6825525 | ||

Dll4
Genomic Map