siRNA Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019454.3 | NP_062327.2 | 10 | 2318 | ||
| GenBank mRNA | AB043893.1 | - | 10 | 1961 | ||
| AF253469.1 | - | 10 | 2273 | |||
| AF273454.1 | - | 10 | 2336 | |||
| AK004739.1 | - | 10 | 2286 | |||
| AK031699.1 | - | 10 | 2288 | |||
| AK052322.1 | - | 10 | 2320 | |||
| AK165517.1 | - | 10 | 2286 | |||
| AK171717.1 | - | 10 | 2285 | |||
| BC042497.1 | - | 10 | 2273 | 4017786 | ||
| BC049130.1 | - | 10 | 2278 | 6825525 | ||

Dll4
Genomic Map