siRNA Details
| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019456.2 | NP_062329.2 | 5, 6 | 743 | ||
| GenBank mRNA | AF020313.1 | - | 2, 3 | 349 | ||
| AK021331.1 | - | 6, 7 | 622 | |||
| AK040084.1 | - | 5, 6 | 679 | |||
| AK041552.1 | - | 5, 6 | 746 | |||
| AK042467.1 | - | 5, 6 | 681 | |||
| AK079736.1 | - | 5, 6 | 977 | |||
| BC003991.1 | - | 2, 3 | 394 | |||
| BC023110.1 | - | 5, 6 | 632 | 3707478 | ||

Apbb1ip
Genomic Map