siRNA Details
| Gene Symbol: Nphs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019459.2 | NP_062332.2 | 26 | 3457 | ||
| GenBank mRNA | AB513652.1 | - | 26 | 3329 | ||
| AF168466.1 | - | 26 | 3329 | |||
| AF172256.1 | - | 26 | 3457 | |||
| AF191090.1 | - | 26 | 3785 | |||
| AK141081.1 | - | 27 | 3752 | |||

Nphs1
Genomic Map