siRNA Details
| Gene Symbol: Olfr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019475.3 | NP_062348.1 | 1 | 1178 | ||
| GenBank mRNA | BC137911.1 | - | 1 | 1175 | 8860930 | |
| BC137912.1 | - | 1 | 1175 | 8860931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr157| Gene Symbol: Olfr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019475.3 | NP_062348.1 | 1 | 1178 | ||
| GenBank mRNA | BC137911.1 | - | 1 | 1175 | 8860930 | |
| BC137912.1 | - | 1 | 1175 | 8860931 | ||