siRNA Details
| Gene Symbol: Pqbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252528.1 | NP_001239457.1 | 4 | 294 | ||
| NM_001252529.1 | NP_001239458.1 | 4 | 360 | |||
| NM_019478.4 | NP_062351.2 | 4 | 489 | |||
| GenBank mRNA | AJ250406.1 | - | 3 | 240 | ||
| AK077652.1 | - | 4 | 319 | |||
| BC009657.1 | - | 3 | 262 | 3484619 | ||
| BC051673.1 | - | 4 | 299 | 30015428 | ||
| BC157976.1 | - | 4 | 394 | 9088137 | ||
| CJ143105.1 | - | 4 | 295 | |||
| CN718083.1 | - | 3 | 274 | |||

Pqbp1
Genomic Map