siRNA Details
| Gene Symbol: Ebag9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019480.4 | NP_062353.3 | 7 | 1015 | ||
| GenBank mRNA | AK004366.1 | - | 6 | 723 | ||
| AK159499.1 | - | 7 | 1065 | |||
| AK160149.1 | - | 7 | 1184 | |||
| AY009091.1 | - | 7 | 1019 | |||
| BC026717.1 | - | 7 | 770 | |||
| BC108397.1 | - | 7 | 1015 | 6329278 | ||
| Gene Symbol: Jpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | JN410359.1 | - | 850 | |||
| JN410360.1 | - | 720 | ||||
| JN410361.1 | - | 757 | ||||

Ebag9
Genomic Map