siRNA Details
| Gene Symbol: Hebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019487.3 | NP_062360.1 | 4 | 1063 | ||
| GenBank mRNA | AF117614.1 | - | 4 | 993 | ||
| AK078130.1 | - | 4 | 1063 | |||
| BC006898.1 | - | 4 | 756 | 3599858 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hebp2| Gene Symbol: Hebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019487.3 | NP_062360.1 | 4 | 1063 | ||
| GenBank mRNA | AF117614.1 | - | 4 | 993 | ||
| AK078130.1 | - | 4 | 1063 | |||
| BC006898.1 | - | 4 | 756 | 3599858 | ||