siRNA Details
| Gene Symbol: Slc2a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019488.4 | NP_062361.1 | 10 | 1480 | ||
| GenBank mRNA | AF232061.2 | - | 10 | 1437 | ||
| AJ245936.1 | - | 10 | 1457 | |||
| AK081806.1 | - | 10 | 1497 | |||
| AK141193.1 | - | 6 | 2520 | |||
| AK155792.1 | - | 10 | 1510 | |||
| AK170319.1 | - | 10 | 1493 | |||
| BC021758.1 | - | 6 | 806 | |||
| BC090993.1 | - | 10 | 1480 | 6751763 | ||
| Y17802.1 | - | 10 | 1462 | |||

Slc2a8
Genomic Map