siRNA Details
| Gene Symbol: Ramp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019511.3 | NP_062384.1 | 3 | 955 | ||
| GenBank mRNA | AF146524.1 | - | 3 | 956 | ||
| AF209907.1 | - | 3 | 915 | |||
| AJ250491.1 | - | 3 | 956 | |||
| AK005513.1 | - | 4 | 1071 | |||
| BC024765.1 | - | 3 | 951 | 5364778 | ||

Ramp3
Genomic Map