siRNA Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019517.4 | NP_062390.3 | 5 | 851 | ||
| GenBank mRNA | AF216310.1 | - | 5 | 802 | ||
| AK052309.1 | - | 5 | 838 | |||
| AK078770.1 | - | 5 | 818 | |||
| AK165036.1 | - | 5 | 787 | |||
| AK183522.1 | - | 2 | 153 | |||
| AK184226.1 | - | 2 | 91 | |||
| AK185329.1 | - | 2 | 91 | |||
| BC120773.1 | - | 5 | 817 | 40129696 | ||
| BC131947.1 | - | 5 | 817 | 40130224 | ||

Bace2
Genomic Map