siRNA Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019517.4 | NP_062390.3 | 9 | 1934 | ||
| GenBank mRNA | AK078770.1 | - | 9 | 1901 | ||
| BC120773.1 | - | 9 | 1900 | 40129696 | ||
| BC131947.1 | - | 9 | 1900 | 40130224 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bace2| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019517.4 | NP_062390.3 | 9 | 1934 | ||
| GenBank mRNA | AK078770.1 | - | 9 | 1901 | ||
| BC120773.1 | - | 9 | 1900 | 40129696 | ||
| BC131947.1 | - | 9 | 1900 | 40130224 | ||