siRNA Details
| Gene Symbol: Grasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019518.3 | NP_062391.3 | 2, 3 | 395 | ||
| GenBank mRNA | AB041603.1 | - | 2, 3 | 393 | ||
| AF236099.1 | - | 2, 3 | 358 | |||
| BC046307.1 | - | 2, 3 | 363 | 6306542 | ||
| BY730445.1 | - | 2, 3 | 414 | |||

Grasp
Genomic Map