siRNA Details
| Gene Symbol: Cts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019539.3 | NP_062412.1 | 2 | 334 | ||
| GenBank mRNA | AF250837.1 | - | 1 | 121 | ||
| AK017466.1 | - | 2 | 360 | |||
| AY014779.1 | - | 2 | 194 | |||
| BC050198.1 | - | 2 | 347 | |||
| BC064740.1 | - | 2 | 334 | 30018572 | ||

Cts7
Genomic Map