siRNA Details
| Gene Symbol: Hao2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019545.4 | NP_062418.3 | 3, 4 | 319 | ||
| GenBank mRNA | AF272947.1 | - | 3, 4 | 315 | ||
| AJ251820.1 | - | 1, 2 | 129 | |||
| AK018684.1 | - | 3, 4 | 307 | |||
| AK078908.1 | - | 3, 4 | 309 | |||
| AK143991.1 | - | 3, 4 | 310 | |||
| BC027754.1 | - | 3, 4 | 351 | 5064100 | ||

Hao2
Genomic Map