siRNA Details
| Gene Symbol: LMAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006816.2 | NP_006807.1 | 3 | 615 | ||
| GenBank mRNA | AK222609.1 | - | 3 | 382 | ||
| AK292099.1 | - | 3 | 392 | |||
| AK301607.1 | - | 4 | 299 | |||
| AK308027.1 | - | 3 | 526 | |||
| BC017263.2 | - | 3 | 395 | 4135210 | ||
| U10362.1 | - | 3 | 378 | |||

LMAN2
Genomic Map