siRNA Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159866.1 | NP_001153338.1 | 2 | 241 | ||
| NM_019562.2 | NP_062508.2 | 2 | 241 | |||
| GenBank mRNA | AF148447.1 | - | 2 | 184 | ||
| AF175903.1 | - | 2 | 193 | |||
| AK007860.1 | - | 1 | 55 | |||
| AK011117.1 | - | 2 | 208 | |||
| AK017925.1 | - | 2 | 239 | |||
| BC006891.1 | - | 2 | 194 | 2650799 | ||
| BY279731.1 | - | 2 | 241 | |||

Uchl5
Genomic Map