siRNA Details
| Gene Symbol: Patz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253690.1 | NP_001240619.1 | 1 | 754 | ||
| NM_001253691.1 | NP_001240620.1 | 1 | 754 | |||
| NM_019574.3 | NP_062520.1 | 1 | 754 | |||
| GenBank mRNA | AB029397.1 | - | 1 | 754 | ||
| AK166189.1 | - | 1 | 875 | |||
| AK185454.1 | - | 1 | 118 | |||
| BC043035.1 | - | 1 | 772 | 5697155 | ||
| BC072649.1 | - | 2 | 1816 | |||

Patz1
Genomic Map