siRNA Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019575.4 | NP_062521.1 | 7 | 1756 | ||
| GenBank mRNA | AF224721.1 | - | 7 | 1731 | ||
| AK019112.1 | - | 7 | 1762 | |||
| AK154094.1 | - | 7 | 1747 | |||
| AK160755.1 | - | 7 | 1747 | |||
| AK168554.1 | - | 7 | 1742 | |||
| BC018215.1 | - | 7 | 1725 | 3965223 | ||
| BC054104.2 | - | 7 | 1750 | 30021104 | ||

Scamp4
Genomic Map