siRNA Details
| Gene Symbol: IFI44L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006820.3 | NP_006811.2 | 2 | 636 | ||
| XM_011540539.2 | XP_011538841.1 | 2 | 474 | |||
| GenBank mRNA | AB000115.1 | - | 2 | 560 | ||
| AK223087.1 | - | 2 | 570 | |||
| AK225442.1 | - | 2 | 537 | |||
| AK312272.1 | - | 2 | 543 | |||
| AL832618.2 | - | 1 | 2682 | |||
| BC015932.2 | - | 2 | 542 | 3909331 | ||

IFI44L
Genomic Map