siRNA Details
| Gene Symbol: Ube2j1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019586.3 | NP_062532.2 | 3 | 383 | ||
| GenBank mRNA | AJ245899.1 | - | 3 | 317 | ||
| AK003050.1 | - | 3 | 315 | |||
| AK031669.1 | - | 3 | 271 | |||
| AK149985.1 | - | 3 | 294 | |||
| AK160428.1 | - | 3 | 389 | |||
| BC024623.1 | - | 3 | 321 | 4217620 | ||
| BY707310.1 | - | 3 | 383 | |||

Ube2j1
Genomic Map