siRNA Details
| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019631.3 | NP_062605.2 | 2 | 472 | ||
| GenBank mRNA | AK049837.1 | - | 2 | 473 | ||
| AK081478.1 | - | 2 | 476 | |||
| BC064715.1 | - | 2 | 475 | 30246737 | ||
| M32486.1 | - | 2 | 172 | |||

Tmem45a
Genomic Map