siRNA Details
| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019631.3 | NP_062605.2 | 3 | 671 | ||
| GenBank mRNA | AK049837.1 | - | 3 | 672 | ||
| AK081478.1 | - | 3 | 675 | |||
| M32486.1 | - | 3 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem45a| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019631.3 | NP_062605.2 | 3 | 671 | ||
| GenBank mRNA | AK049837.1 | - | 3 | 672 | ||
| AK081478.1 | - | 3 | 675 | |||
| M32486.1 | - | 3 | 371 | |||