siRNA Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019637.3 | NP_062611.2 | 10 | 864 | ||
| GenBank mRNA | AK002822.1 | - | 806 | |||
| AK030122.1 | - | 10 | 864 | |||
| AK042741.1 | - | 11 | 919 | |||
| AK046970.1 | - | 10 | 841 | |||
| AK154136.1 | - | 10 | 959 | |||
| AK166043.1 | - | 10 | 847 | |||
| BC038608.1 | - | 10 | 814 | 5012303 | ||
| BC096674.1 | - | 9 | 774 | 5144003 | ||
| U34973.1 | - | 11 | 968 | |||

Styx
Genomic Map