siRNA Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019637.3 | NP_062611.2 | 11 | 1916 | ||
| GenBank mRNA | AK014193.1 | - | 1 | 269 | ||
| AK030122.1 | - | 11 | 1916 | |||
| AK031186.1 | - | 1 | 978 | |||
| AK046970.1 | - | 11 | 1893 | |||
| AK154136.1 | - | 11 | 2011 | |||
| AK166043.1 | - | 11 | 1898 | |||

Styx
Genomic Map