siRNA Details
| Gene Symbol: Clptm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019649.2 | NP_062623.2 | 11 | 1418 | ||
| GenBank mRNA | AK148080.1 | - | 11 | 1400 | ||
| AK152410.1 | - | 11 | 1408 | |||
| AK152809.1 | - | 11 | 1408 | |||
| AK165742.1 | - | 10 | 2112 | |||
| AK171901.1 | - | 11 | 1409 | |||
| AK220120.1 | - | 1 | 64 | |||
| BC022172.1 | - | 11 | 1409 | 4193279 | ||
| D67067.1 | - | 11 | 1405 | |||

Clptm1
Genomic Map