siRNA Details
| Gene Symbol: Clptm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019649.2 | NP_062623.2 | 14 | 1821 | ||
| GenBank mRNA | AK148080.1 | - | 14 | 1803 | ||
| AK165742.1 | - | 13 | 2515 | |||
| AK171901.1 | - | 13 | 2466 | |||
| BC018389.1 | - | 4 | 407 | |||
| BC022172.1 | - | 14 | 1812 | 4193279 | ||
| D67067.1 | - | 14 | 1807 | |||

Clptm1
Genomic Map