siRNA Details
| Gene Symbol: Shoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168505.1 | NP_001161977.1 | 2 | 564 | ||
| NM_019658.6 | NP_062632.2 | 2 | 605 | |||
| GenBank mRNA | AF068921.1 | - | 1 | 240 | ||
| AK146447.1 | - | 2 | 602 | |||
| BC026364.1 | - | 1 | 471 | 4014650 | ||
| BC049775.1 | - | 1 | 470 | 6515905 | ||
| BC083060.1 | - | 2 | 564 | 5356514 | ||

Shoc2
Genomic Map