siRNA Details
| Gene Symbol: Arl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019665.3 | NP_062639.3 | 8 | 1301 | ||
| GenBank mRNA | AF031903.1 | - | 8 | 1019 | ||
| AK003778.1 | - | 8 | 1129 | |||
| AK008897.1 | - | 1 | 1271 | |||
| AK152980.1 | - | 8 | 1106 | |||
| AK158832.1 | - | 9 | 1367 | |||
| AK160666.1 | - | 8 | 1120 | |||
| BC018497.1 | - | 8 | 1121 | 3484868 | ||

Arl6
Genomic Map