siRNA Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019670.1 | NP_062644.1 | 15 | 1569 | ||
| GenBank mRNA | AF094519.1 | - | 15 | 1569 | ||
| AK137073.1 | - | 14 | 1658 | |||
| AK138694.1 | - | 15 | 1599 | |||
| AK161737.1 | - | 14 | 1587 | |||
| AK166882.1 | - | 15 | 1625 | |||
| AK220493.1 | - | 15 | 1633 | |||
| BC003250.1 | - | 14 | 1603 | |||
| BC028920.1 | - | 8 | 870 | 4008236 | ||
| BC085191.1 | - | 15 | 1679 | 30693873 | ||
| BC086779.1 | - | 15 | 1644 | |||

Diap3
Genomic Map