siRNA Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047159.2 | NP_001040624.1 | 3 | 497 | ||
NM_019671.3 | NP_062645.2 | 5 | 618 | |||
GenBank mRNA | AF094520.1 | - | 5 | 514 | ||
AJ010045.1 | - | 3 | 460 | |||
AK082128.1 | - | 3 | 494 | |||
AK088632.1 | - | 3 | 461 | |||
AK139921.1 | - | 3 | 461 | |||
BC004699.1 | - | 3 | 471 | 3499258 | ||
CN702942.1 | - | 3 | 358 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
