siRNA Details
| Gene Symbol: Ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019674.3 | NP_062648.1 | 5 | 377 | ||
| GenBank mRNA | AF088911.1 | - | 5 | 402 | ||
| AK027923.1 | - | 3 | 156 | |||
| AK144244.1 | - | 4 | 304 | |||
| BC001993.1 | - | 5 | 377 | 3491568 | ||
| U79747.1 | - | 1 | 29 | |||

Ppp4c
Genomic Map