siRNA Details
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019699.1 | NP_062673.1 | 11 | 1259 | ||
| GenBank mRNA | AF126798.1 | - | 11 | 1259 | ||
| AK083282.1 | - | 1226 | ||||
| AK143524.1 | - | 1277 | ||||
| BC057189.1 | - | 11 | 1247 | 5137343 | ||
| HQ264057.1 | - | 11 | 1152 | |||
| HQ264058.1 | - | 11 | 1152 | |||

Fads2
Genomic Map