siRNA Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291071.1 | NP_001278000.1 | 4 | 395 | ||
| NM_019703.4 | NP_062677.1 | 4 | 395 | |||
| GenBank mRNA | AF123533.1 | - | 4 | 360 | ||
| AK004790.1 | - | 4 | 350 | |||
| AK020367.1 | - | 4 | 350 | |||
| AK049841.1 | - | 4 | 349 | |||
| AK054193.1 | - | 4 | 350 | |||
| AK076803.1 | - | 4 | 350 | |||
| AK078254.1 | - | 4 | 350 | |||
| AK098162.1 | - | 4 | 349 | |||
| AK152670.1 | - | 4 | 350 | |||
| AK165422.1 | - | 4 | 349 | |||
| AK165425.1 | - | 4 | 349 | |||
| AK170624.1 | - | 4 | 350 | |||
| AK171062.1 | - | 4 | 350 | |||
| AY779275.1 | - | 4 | 312 | |||
| AY779276.1 | - | 4 | 312 | |||
| BC006926.1 | - | 4 | 334 | 2655383 | ||
| BY243493.1 | - | 4 | 394 | |||
| CT010268.1 | - | 4 | 303 | |||
| Y19008.1 | - | 4 | 352 | |||

Pfkp
Genomic Map