siRNA Details
| Gene Symbol: ILVBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006844.4 | NP_006835.2 | 16 | 2217 | ||
| XM_005259717.4 | XP_005259774.1 | 16 | 2364 | |||
| XM_011527651.2 | XP_011525953.1 | 15 | 2196 | |||
| GenBank mRNA | AB209065.1 | - | 10 | 4144 | ||
| BC000109.1 | - | 5 | 839 | |||
| BC011722.2 | - | 16 | 2184 | 4300207 | ||
| BC011761.2 | - | 16 | 2110 | 3622453 | ||
| U61263.1 | - | 16 | 2092 | |||

ILVBL
Genomic Map