siRNA Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285805.1 | NP_001272734.1 | 10 | 1307 | ||
| NM_019778.2 | NP_062752.2 | 8 | 1254 | |||
| NM_181058.1 | NP_851401.1 | 6 | 1032 | |||
| GenBank mRNA | AB221632.1 | - | 2 | 719 | ||
| AF185576.1 | - | 6 | 1077 | |||
| AF194030.1 | - | 4 | 869 | |||
| AK005028.2 | - | 6 | 1033 | |||
| AY028963.1 | - | 3 | 836 | |||
| BC031114.1 | - | 7 | 1140 | 4971586 | ||
| BC056446.1 | - | 10 | 1279 | 5703319 | ||

Zbtb20
Genomic Map