siRNA Details
| Gene Symbol: CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254750.1 | NP_001241679.1 | 9, 10 | 1715 | ||
| NM_001254751.1 | NP_001241680.1 | 9, 10 | 1688 | |||
| NM_006725.4 | NP_006716.3 | 10, 11 | 1811 | |||
| NR_045638.1 | 10, 11 | 1791 | ||||
| XM_006718738.1 | XP_006718801.1 | 9, 10 | 1715 | |||
| XM_006718739.1 | XP_006718802.1 | 9, 10 | 1688 | |||
| XM_006718740.1 | XP_006718803.1 | 9, 10 | 1688 | |||
| XM_006718741.1 | XP_006718804.1 | 8, 9 | 1592 | |||
| XM_011545360.1 | XP_011543662.1 | 10, 11 | 1811 | |||
| XM_011545362.1 | XP_011543664.1 | 9, 10 | 1508 | |||
| GenBank mRNA | AK128419.1 | - | 11, 12 | 1715 | ||
| AK292704.1 | - | 10, 11 | 1687 | |||
| AK312749.1 | - | 10, 11 | 1772 | |||
| BC033755.1 | - | 10, 11 | 1727 | 5213263 | ||
| BC078669.1 | - | 10, 11 | 1750 | 6302947 | ||
| U66142.1 | - | 10, 11 | 1586 | |||
| U66145.1 | - | 9, 10 | 1490 | |||
| U66146.1 | - | 9, 10 | 1463 | |||
| X60992.1 | - | 10, 11 | 1686 | |||

CD6
Genomic Map