siRNA Details
| Gene Symbol: Cyp3a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019792.2 | NP_062766.2 | 2 | 206 | ||
| GenBank mRNA | AF204959.1 | - | 2 | 205 | ||
| AK149541.1 | - | 2 | 208 | |||
| BC028855.1 | - | 2 | 197 | 5124151 | ||
| BY099368.1 | - | 2 | 208 | |||
| Y11995.1 | - | 2 | 208 | |||

Cyp3a25
Genomic Map