siRNA Details
| Gene Symbol: Vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019806.5 | NP_062780.2 | 5, 6 | 703 | ||
| GenBank mRNA | AF115504.1 | - | 5, 6 | 581 | ||
| AK050632.1 | - | 5, 6 | 652 | |||
| AK081908.1 | - | 5, 6 | 632 | |||
| AK160925.1 | - | 5, 6 | 546 | |||
| AK172080.1 | - | 5, 6 | 476 | |||
| BC048231.1 | - | 5, 6 | 703 | 6313869 | ||
| BC057123.1 | - | 5, 6 | 692 | 6411688 | ||

Vapb
Genomic Map