siRNA Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019816.1 | NP_062790.1 | 2 | 324 | ||
| GenBank mRNA | AF222801.1 | - | 2 | 324 | ||
| AK077789.1 | - | 2 | 348 | |||
| AK141334.1 | - | 2 | 322 | |||
| AY306199.1 | - | 2 | 205 | |||
| BC025080.1 | - | 2 | 311 | 2648117 | ||

Aatf
Genomic Map