siRNA Details
| Gene Symbol: Ncoa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242558.1 | NP_001229487.1 | 13 | 3841 | ||
| NM_019825.3 | NP_062799.2 | 14 | 6817 | |||
| GenBank mRNA | AF216186.1 | - | 13 | 6702 | ||
| AK129079.1 | - | 7 | 3976 | |||
| AK136585.1 | - | 4 | 1589 | |||
| AK161598.1 | - | 2 | 1617 | |||
| BC083187.1 | - | 14 | 6817 | 30544739 | ||

Ncoa6
Genomic Map