siRNA Details
| Gene Symbol: Fam69b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019833.3 | NP_062807.2 | 5 | 1436 | ||
| GenBank mRNA | AB030186.1 | - | 5 | 1430 | ||
| AF332189.1 | - | 5 | 1430 | |||
| AK045880.1 | - | 5 | 1437 | |||
| AK050584.1 | - | 5 | 1351 | |||
| AK171112.1 | - | 4 | 5296 | |||
| BC051954.1 | - | 4 | 1208 | 5714977 | ||
| BC060081.1 | - | 4 | 1208 | 5714977 | ||
| BC116751.1 | - | 5 | 1223 | 40126070 | ||
| BC116779.1 | - | 5 | 1223 | |||

Fam69b
Genomic Map