siRNA Details
| Gene Symbol: Rbm14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019869.3 | NP_063922.2 | 1, 2 | 461 | ||
| GenBank mRNA | AK004928.1 | - | 1, 2 | 431 | ||
| AK050945.1 | - | 1, 2 | 463 | |||
| AK088201.1 | - | 1, 2 | 427 | |||
| AK153230.1 | - | 1, 2 | 436 | |||
| AK167503.1 | - | 1, 2 | 440 | |||
| BC010294.1 | - | 1, 2 | 412 | 3153900 | ||

Rbm14
Genomic Map