siRNA Details
| Gene Symbol: Dnajb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019874.3 | NP_063927.1 | 3 | 1715 | ||
| GenBank mRNA | AK004300.1 | - | 1 | 349 | ||
| AK158972.1 | - | 4 | 2015 | |||
| BC027782.1 | - | 3 | 4457 | |||
| BC057087.1 | - | 3 | 1715 | 5695243 | ||

Dnajb5
Genomic Map